MilliporeSigma
  • Intranasal administration of a single dose of a candidate live attenuated vaccine derived from an NSP16-deficient SARS-CoV-2 strain confers sterilizing immunity in animals.
Intranasal administration of a single dose of a candidate live attenuated vaccine derived from an NSP16-deficient SARS-CoV-2 strain confers sterilizing immunity in animals.
Cellular & molecular immunology (2022-03-31)
Zi-Wei Ye, Chon Phin Ong, Kaiming Tang, Yilan Fan, Cuiting Luo, Runhong Zhou, Peng Luo, Yun Cheng, Victor Sebastien Gray, Pui Wang, Hin Chu, Jasper Fuk-Woo Chan, Kelvin Kai-Wang To, Honglin Chen, Zhiwei Chen, Kwok-Yung Yuen, Guang Sheng Ling, Shuofeng Yuan, Dong-Yan Jin
ABSTRACT
Live attenuated vaccines might elicit mucosal and sterilizing immunity against SARS-CoV-2 that the existing mRNA, adenoviral vector and inactivated vaccines fail to induce. Here, we describe a candidate live attenuated vaccine strain of SARS-CoV-2 in which the NSP16 gene, which encodes 2'-O-methyltransferase, is catalytically disrupted by a point mutation. This virus, designated d16, was severely attenuated in hamsters and transgenic mice, causing only asymptomatic and nonpathogenic infection. A single dose of d16 administered intranasally resulted in sterilizing immunity in both the upper and lower respiratory tracts of hamsters, thus preventing viral spread in a contact-based transmission model. It also robustly stimulated humoral and cell-mediated immune responses, thus conferring full protection against lethal challenge with SARS-CoV-2 in a transgenic mouse model. The neutralizing antibodies elicited by d16 effectively cross-reacted with several SARS-CoV-2 variants. Secretory immunoglobulin A was detected in the blood and nasal wash of vaccinated mice. Our work provides proof-of-principle evidence for harnessing NSP16-deficient SARS-CoV-2 for the development of live attenuated vaccines and paves the way for further preclinical studies of d16 as a prototypic vaccine strain, to which new features might be introduced to improve safety, transmissibility, immunogenicity and efficacy.
Sigma-Aldrich
Anti-Mouse IgA antibody, Rabbit monoclonal, recombinant, expressed in HEK 293 cells, clone RM220, purified immunoglobulin
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) FITC conjugate, 1.0 mg/mL, Chemicon®