MilliporeSigma
  • Sex Differences in Protein Kinase A Signaling of the Latent Postoperative Pain Sensitization That Is Masked by Kappa Opioid Receptors in the Spinal Cord.
Sex Differences in Protein Kinase A Signaling of the Latent Postoperative Pain Sensitization That Is Masked by Kappa Opioid Receptors in the Spinal Cord.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2021-09-18)
Paramita Basu, Lilian Custodio-Patsey, Pranav Prasoon, Bret N Smith, Bradley K Taylor
ABSTRACT
Latent sensitization (LS) of pain engages pronociceptive signaling pathways in the dorsal horn that include NMDA receptor (NMDAR)→adenylyl cyclase-1 (AC1)→protein kinase A (PKA), and exchange proteins directly activated by cyclic AMP (Epacs). To determine whether these pathways operate similarly between males and females or are under the inhibitory control of spinal κ opioid receptors (KOR), we allowed hyperalgesia to resolve after plantar incision and then blocked KOR with intrathecal administration of LY2456302, which reinstated hyperalgesia and facilitated touch-evoked immunoreactivity of phosphorylated extracellular signal-regulated kinase (pERK) in neurons (NeuN) but not astrocytes (GFAPs) nor microglia (Iba1). LY2456302 reinstated hyperalgesia even when administered 13 months later, indicating that chronic postoperative pain vulnerability persists for over a year in a latent state of remission. In both sexes, intrathecal MK-801 (an NMDAR competitive antagonist) prevented LY2456302-evoked reinstatement of hyperalgesia as did AC1 gene deletion or the AC1 inhibitor NB001. NB001 also prevented stimulus-evoked pERK. In both sexes, the Epac inhibitor ESI-09 prevented reinstatement, whereas the Epac activator 8-CPT reinstated hyperalgesia. By contrast, the PKA inhibitor H89 prevented reinstatement only in male mice, whereas the PKA activator 6-bnz-cAMP itself evoked reinstatement at all doses tested (3-30 nmol, i.t.). In neither sex did incision change gene expression of KOR, GluN1, PKA, or Epac1 in dorsal horn. We conclude that sustained KOR signaling inhibits spinal PKA-dependent mechanisms that drive postoperative LS in a sex-dependent manner. Our findings support the development of AC1, PKA, and Epac inhibitors toward a new pharmacotherapy for chronic postoperative pain.SIGNIFICANCE STATEMENT Because of neural mechanisms that are not well understood, men and women respond differently to treatments for chronic pain. We report that surgical incision recruits a pronociceptive latent pain sensitization that persisted for over a year and was kept in check by the sustained analgesic activity of κ opioid receptors. NMDAR→AC1→cAMP→Epac signaling pathways in the dorsal horn of the spinal cord maintain latent sensitization in both males and females; however, only males recruit a PKA-dependent mechanism. This work presents a novel male-specific mechanism for the promotion of chronic postoperative pain.
Sigma-Aldrich
Anti-AIF1/IBA1 (Isoforms 1 and 3) antibody produced in goat, affinity isolated antibody, buffered aqueous solution