MilliporeSigma
  • The role of oxytocin release in the paraventricular nucleus in the control of maternal behaviour in the sheep.
The role of oxytocin release in the paraventricular nucleus in the control of maternal behaviour in the sheep.
Journal of neuroendocrinology (1996-03-01)
A P Da Costa, R G Guevara-Guzman, S Ohkura, J A Goode, K M Kendrick
ABSTRACT
Oxytocin (OT) release within the brain is thought to play a major role in inducing maternal behaviour in a number of mammalian species but little is known about the sites of release which are important in this respect. We have investigated whether the paraventricular nucleus of the hypothalamus (PVN) is a site of OT action on maternal behaviour in the sheep. In vivo microdialysis and retrodialysis was used to determine whether OT is released in the region of the PVN during the post-partum induction of maternal behaviour and if its release at this site can stimulate maternal behaviour in non-pregnant animals. In vivo sampling showed that OT concentrations increased significantly in the region of PVN at birth. When OT was retrodialysed bilaterally into the PVN (1 or 10 microM) of multiparous ewes treated with progesterone and oestradiol to stimulate lactation, maternal behaviour was induced in a significant number of animals (1 microM, 6/8 and 10 microM, 5/8) compared with controls (0/8 ewes). Similar infusions of the ring structure of OT, tocinoic acid (TOC-10 microM), also induced maternal behaviour in a significant proportion of animals (5/6 ewes) as did intracerebroventricular (ICV) OT (6/8 ewes) and artificial stimulation of the vagina and cervix (VCS, 8/9 ewes). On the other hand, vasopressin (AVP) 1 microM did not induce maternal behaviour in any ewes and a 10 microM dose only induced it in 2/8 animals. The neurochemical changes accompanying the above treatments were also investigated. Noradrenaline concentrations increased in the PVN after the retrodialysis administration of OT 1 microM and 10 microM, TOC 10 microM and AVP 1 microM, OT ICV and VCS. Dopamine concentrations were also increased by OT 10 microM, TOC 10 microM, AVP 1microM and OT ICV. Aspartate and glutamate concentrations were significantly reduced by retrodialysis infusions of OT 1 microM and AVP 1 and 10 microM but not by any other treatment. Finally, the retrodialysis infusion of OT and TOC, as well as ICV OT, significantly increased plasma OT release whereas AVP infusions did not. These results provide evidence that OT is released in the PVN during parturition and is important for the induction of maternal behaviour. It seems probable that OT release at this site has a positive feedback effect on both parvocellular and magnocellular OT neurons to facilitate co-ordinated OT release both in central OT terminal regions (to facilitate maternal behaviour) and peripherally into the blood (to facilitate uterine contractions/milk let down). The potential functional roles for the actions of OT on monoamine and amino acid transmitter release in the PVN are discussed.