MilliporeSigma
  • Structural basis of malaria transmission blockade by a monoclonal antibody to gamete fusogen HAP2.
Structural basis of malaria transmission blockade by a monoclonal antibody to gamete fusogen HAP2.
eLife (2021-12-24)
Juan Feng, Xianchi Dong, Adam DeCosta, Yang Su, Fiona Angrisano, Katarzyna A Sala, Andrew M Blagborough, Chafen Lu, Timothy A Springer
ABSTRACT
HAP2 is a transmembrane gamete fusogen found in multiple eukaryotic kingdoms and is structurally homologous to viral class II fusogens. Studies in Plasmodium have suggested that HAP2 is an attractive target for vaccines that block transmission of malaria. HAP2 has three extracellular domains, arranged in the order D2, D1, and D3. Here, we report monoclonal antibodies against the D3 fragment of Plasmodium berghei HAP2 and crystal structures of D3 in complex with Fab fragments of two of these antibodies, one of which blocks fertilization of Plasmodium berghei in vitro and transmission of malaria in mosquitoes. We also show how this Fab binds the complete HAP2 ectodomain with electron microscopy. The two antibodies cross-react with HAP2 among multiple plasmodial species. Our characterization of the Plasmodium D3 structure, HAP2 ectodomain architecture, and mechanism of inhibition provide insights for the development of a vaccine to block malaria transmission.
Sigma-Aldrich
IgG2a, Kappa from murine myeloma, clone UPC 10, ascites fluid, lyophilized powder