MilliporeSigma
  • A single-cell atlas of the normal and malformed human brain vasculature.
A single-cell atlas of the normal and malformed human brain vasculature.
Science (New York, N.Y.) (2022-01-28)
Ethan A Winkler, Chang N Kim, Jayden M Ross, Joseph H Garcia, Eugene Gil, Irene Oh, Lindsay Q Chen, David Wu, Joshua S Catapano, Kunal Raygor, Kazim Narsinh, Helen Kim, Shantel Weinsheimer, Daniel L Cooke, Brian P Walcott, Michael T Lawton, Nalin Gupta, Berislav V Zlokovic, Edward F Chang, Adib A Abla, Daniel A Lim, Tomasz J Nowakowski
ABSTRACT
Cerebrovascular diseases are a leading cause of death and neurologic disability. Further understanding of disease mechanisms and therapeutic strategies requires a deeper knowledge of cerebrovascular cells in humans. We profiled transcriptomes of 181,388 cells to define a cell atlas of the adult human cerebrovasculature, including endothelial cell molecular signatures with arteriovenous segmentation and expanded perivascular cell diversity. By leveraging this reference, we investigated cellular and molecular perturbations in brain arteriovenous malformations, which are a leading cause of stroke in young people, and identified pathologic endothelial transformations with abnormal vascular patterning and the ontology of vascularly derived inflammation. We illustrate the interplay between vascular and immune cells that contributes to brain hemorrhage and catalog opportunities for targeting angiogenic and inflammatory programs in vascular malformations.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-PLVAP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Osteopontin human, recombinant, expressed in HEK 293 cells, ≥97% (SDS-PAGE), ≥97% (HPLC), suitable for cell culture
Sigma-Aldrich
Anti-P2RY12 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution