MilliporeSigma
  • NBTI attenuates neuroinflammation and apoptosis partly by ENT1/NLRP3/Bcl2 pathway after subarachnoid hemorrhage in rats.
NBTI attenuates neuroinflammation and apoptosis partly by ENT1/NLRP3/Bcl2 pathway after subarachnoid hemorrhage in rats.
Neuroreport (2021-11-01)
Xiaowei Chen, Xiaocheng Luo, Hang Hu, Qianghua Xu
ABSTRACT
Neuroinflammation and apoptosis are two key factors contributing to early brain injury (EBI) after subarachnoid hemorrhage (SAH) and are strongly associated with a poor prognosis. Recently, equilibrative nucleoside transporter 1 (ENT1) was emerged to accelerate the severity of inflammation and cell apoptosis in several nervous system diseases, including cerebral ischemia, neurodegeneration and epilepsy. However, no study has yet elaborated the expression levels and effects of ENT1 in EBI after SAH. Sprague-Dawley rats were subjected to SAH by endovascular perforation. Nitrobenzylthioinosine (NBTI) was intranasally administered at 0.5 h after SAH. The protein expression levels of ENT1, NLRP3, Bcl2, Bax, ACS, Caspase-1, IL-1 were detected by western blot. The modified Garcia score and beam balance score were employed to evaluate the neurologic function of rats following SAH. In addition, hematoxylin-eosin, fluoro-jade C and TdT-mediated dUTP nick-end labeling staining were then used to evaluate brain tissue damage and neuronal apoptosis. Analysis indicated that endogenous levels of ENT1 were significantly upregulated at 24-hour post-SAH, accompanied by NLRP3 inflammasome activation and Bcl2 decline. The administration of NBTI, an inhibitor of ENT1, at a dose of 15 mg/kg, ameliorated neurologic deficits and morphologic lesions at both 24 and 72 h after SAH. Moreover, ENT1 inhibition efficiently mitigated neuronal degeneration and cell apoptosis. In addition, NBTI at 15 mg/kg observably increased Bcl2 content and decreased Bax level. Furthermore, suppression of ENT1 notably reduced the expression levels of NLRP3, apoptosis associated speck like protein containing CARD, caspase-1 and IL-1β. NBTI relieved SAH-induced EBI partly through ENT1/NLRP3/Bcl2 pathway.
Sigma-Aldrich
Anti-Caspase 1 Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.