MilliporeSigma
  • DNA methylation-based age acceleration observed in IDH wild-type glioblastoma is associated with better outcome-including in elderly patients.
DNA methylation-based age acceleration observed in IDH wild-type glioblastoma is associated with better outcome-including in elderly patients.
Acta neuropathologica communications (2022-03-26)
Pierre Bady, Christine Marosi, Michael Weller, Bjørn H Grønberg, Henrik Schultz, Martin J B Taphoorn, Johanna M M Gijtenbeek, Martin J van den Bent, Andreas von Deimling, Roger Stupp, Annika Malmström, Monika E Hegi
ABSTRACT
Elderly patients represent a growing proportion of individuals with glioblastoma, who however, are often excluded from clinical trials owing to poor expected prognosis. We aimed at identifying age-related molecular differences that would justify and guide distinct treatment decisions in elderly glioblastoma patients. The combined DNA methylome (450 k) of four IDH wild-type glioblastoma datasets, comprising two clinical trial cohorts, was interrogated for differences based on the patients' age, DNA methylation (DNAm) age acceleration (DNAm age "Horvath-clock" minus patient age), DNA methylation-based tumor classification (Heidelberg), entropy, and functional methylation of DNA damage response (DDR) genes. Age dependent methylation included 19 CpGs (p-value ≤ 0.1, Bonferroni corrected), comprising a CpG located in the ELOVL2 gene that is part of a 13-gene forensic age predictor. Most of the age related CpGs (n = 16) were also associated with age acceleration that itself was associated with a large number of CpGs (n = 50,551). Over 70% age acceleration-associated CpGs (n = 36,348) overlapped with those associated with the DNA methylation based tumor classification (n = 170,759). Gene set enrichment analysis identified associated pathways, providing insights into the biology of DNAm age acceleration and respective commonalities with glioblastoma classification. Functional methylation of several DDR genes, defined as correlation of methylation with gene expression (r ≤ -0.3), was associated with age acceleration (n = 8), tumor classification (n = 12), or both (n = 4), the latter including MGMT. DNAm age acceleration was significantly associated with better outcome in both clinical trial cohorts, whereof one comprised only elderly patients. Multivariate analysis included treatment (RT, RT/TMZ→TMZ; TMZ, RT), MGMT promoter methylation status, and interaction with treatment. In conclusion, DNA methylation features of age acceleration are an integrative part of the methylation-based tumor classification (RTK I, RTK II, MES), while patient age seems hardly reflected in the glioblastoma DNA methylome. We found no molecular evidence justifying other treatments in elderly patients, not owing to frailty or co-morbidities.
Product Description
Millipore
EX-WAX Paraffin-embedded DNA Extraction Kit