MilliporeSigma
  • P300/CBP-associated factor regulates transcription and function of isocitrate dehydrogenase 2 during muscle differentiation.
P300/CBP-associated factor regulates transcription and function of isocitrate dehydrogenase 2 during muscle differentiation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-12-12)
Matteo Savoia, Chiara Cencioni, Mattia Mori, Sandra Atlante, Germana Zaccagnini, Paolo Devanna, Lucia Di Marcotullio, Bruno Botta, Fabio Martelli, Andreas M Zeiher, Alfredo Pontecorvi, Antonella Farsetti, Francesco Spallotta, Carlo Gaetano
ABSTRACT
The epigenetic enzyme p300/CBP-associated factor (PCAF) belongs to the GCN5-related N-acetyltransferase (GNAT) family together with GCN5. Although its transcriptional and post-translational function is well characterized, little is known about its properties as regulator of cell metabolism. Here, we report the mitochondrial localization of PCAF conferred by an 85 aa mitochondrial targeting sequence (MTS) at the N-terminal region of the protein. In mitochondria, one of the PCAF targets is the isocitrate dehydrogenase 2 (IDH2) acetylated at lysine 180. This PCAF-regulated post-translational modification might reduce IDH2 affinity for isocitrate as a result of a conformational shift involving predictively the tyrosine at position 179. Site-directed mutagenesis and functional studies indicate that PCAF regulates IDH2, acting at dual level during myoblast differentiation: at a transcriptional level together with MyoD, and at a post-translational level by direct modification of lysine acetylation in mitochondria. The latter event determines a decrease in IDH2 function with negative consequences on muscle fiber formation in C2C12 cells. Indeed, a MTS-deprived PCAF does not localize into mitochondria, remains enriched into the nucleus, and contributes to a significant increase of muscle-specific gene expression enhancing muscle differentiation. The role of PCAF in mitochondria is a novel finding shedding light on metabolic processes relevant to early muscle precursor differentiation.-Savoia, M., Cencioni, C., Mori, M., Atlante, S., Zaccagnini, G., Devanna, P., Di Marcotullio, L., Botta, B., Martelli, F., Zeiher, A. M., Pontecorvi, A., Farsetti, A., Spallotta, F., Gaetano, C. P300/CBP-associated factor regulates transcription and function of isocitrate dehydrogenase 2 during muscle differentiation.
Sigma-Aldrich
PCAF Protein, Active, 50 µg, PCAF recombinant GST fusion protein, for use in Histone Acetyltransferase Assays. (HAT).