MilliporeSigma
The flavonoid corylin exhibits lifespan extension properties in mouse.
Nature communications (2022-03-11)
Tong-Hong Wang, Wei-Che Tseng, Yann-Lii Leu, Chi-Yuan Chen, Wen-Chih Lee, Ying-Chih Chi, Shu-Fang Cheng, Chun-Yu Lai, Chen-Hsin Kuo, Shu-Ling Yang, Sien-Hung Yang, Jiann-Jong Shen, Chun-Hao Feng, Chih-Ching Wu, Tsong-Long Hwang, Chia-Jen Wang, Shu-Huei Wang, Chin-Chuan Chen
ABSTRACT
In the long history of traditional Chinese medicine, single herbs and complex formulas have been suggested to increase lifespan. However, the identification of single molecules responsible for lifespan extension has been challenging. Here, we collected a list of traditional Chinese medicines with potential longevity properties from pharmacopeias. By utilizing the mother enrichment program, we systematically screened these traditional Chinese medicines and identified a single herb, Psoralea corylifolia, that increases lifespan in Saccharomyces cerevisiae. Next, twenty-two pure compounds were isolated from Psoralea corylifolia. One of the compounds, corylin, was found to extend the replicative lifespan in yeast by targeting the Gtr1 protein. In human umbilical vein endothelial cells, RNA sequencing data showed that corylin ameliorates cellular senescence. We also examined an in vivo mammalian model, and found that corylin extends lifespan in mice fed a high-fat diet. Taken together, these findings suggest that corylin may promote longevity.
Sigma-Aldrich
Anti-GFP, N-terminal antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Corylin, ≥98% (HPLC)