MilliporeSigma
  • Direct reprogramming of mouse and human fibroblasts into multipotent neural stem cells with a single factor.
Direct reprogramming of mouse and human fibroblasts into multipotent neural stem cells with a single factor.
Cell stem cell (2012-06-12)
Karen L Ring, Leslie M Tong, Maureen E Balestra, Robyn Javier, Yaisa Andrews-Zwilling, Gang Li, David Walker, William R Zhang, Anatol C Kreitzer, Yadong Huang
ABSTRACT
The generation of induced pluripotent stem cells (iPSCs) and induced neuronal cells (iNCs) from somatic cells provides new avenues for basic research and potential transplantation therapies for neurological diseases. However, clinical applications must consider the risk of tumor formation by iPSCs and the inability of iNCs to self-renew in culture. Here we report the generation of induced neural stem cells (iNSCs) from mouse and human fibroblasts by direct reprogramming with a single factor, Sox2. iNSCs express NSC markers and resemble wild-type NSCs in their morphology, self-renewal, ability to form neurospheres, and gene expression profiles. Cloned iNSCs differentiate into several types of mature neurons, as well as astrocytes and oligodendrocytes, indicating multipotency. Implanted iNSCs can survive and integrate in mouse brains and, unlike iPSC-derived NSCs, do not generate tumors. Thus, self-renewable and multipotent iNSCs without tumorigenic potential can be generated directly from fibroblasts by reprogramming.
Sigma-Aldrich
Cryopreserved Mouse Cortical Neural Stem Cells, The Cryopreserved Mouse Cortical Neural Stem Cells provides ready-to use primary neural stem cells isolated from the cortices of embryonic day 15-18 (E15-E18) C57/BL6 mice.