MilliporeSigma
  • APOE immunotherapy reduces cerebral amyloid angiopathy and amyloid plaques while improving cerebrovascular function.
APOE immunotherapy reduces cerebral amyloid angiopathy and amyloid plaques while improving cerebrovascular function.
Science translational medicine (2021-02-19)
Monica Xiong, Hong Jiang, Javier Remolina Serrano, Ernesto R Gonzales, Chao Wang, Maud Gratuze, Rosa Hoyle, Nga Bien-Ly, Adam P Silverman, Patrick M Sullivan, Ryan J Watts, Jason D Ulrich, Gregory J Zipfel, David M Holtzman
ABSTRACT
The ε4 allele of the apolipoprotein E (APOE) gene is the strongest genetic risk factor for late-onset Alzheimer's disease (AD) and greatly influences the development of amyloid-β (Aβ) pathology. Our current study investigated the potential therapeutic effects of the anti-human APOE antibody HAE-4, which selectively recognizes human APOE that is co-deposited with Aβ in cerebral amyloid angiopathy (CAA) and parenchymal amyloid pathology. In addition, we tested whether HAE-4 provoked brain hemorrhages, a component of amyloid-related imaging abnormalities (ARIA). ARIA is an adverse effect secondary to treatment with anti-Aβ antibodies that can occur in blood vessels with CAA. We used 5XFAD mice expressing human APOE4 +/+ (5XE4) that have prominent CAA and parenchymal plaque pathology to assess the efficacy of HAE-4 compared to an Aβ antibody that removes parenchymal Aβ but increases ARIA in humans. In chronically treated 5XE4 mice, HAE-4 reduced Aβ deposition including CAA compared to a control antibody, whereas the anti-Aβ antibody had no effect on CAA. Furthermore, the anti-Aβ antibody exacerbated microhemorrhage severity, which highly correlated with reactive astrocytes surrounding CAA. In contrast, HAE-4 did not stimulate microhemorrhages and instead rescued CAA-induced cerebrovascular dysfunction in leptomeningeal arteries in vivo. HAE-4 not only reduced amyloid but also dampened reactive microglial, astrocytic, and proinflammatory-associated genes in the cortex. These results suggest that targeting APOE in the core of both CAA and plaques could ameliorate amyloid pathology while protecting cerebrovascular integrity and function.
Sigma-Aldrich
Anti-PECAM-1 Antibody, clone 2H8, Azide Free, clone 2H8, Chemicon®, from hamster(Armenian)
Sigma-Aldrich
X-34, ≥90% (HPLC)
Sigma-Aldrich
Anti-GFAP Antibody, Biotin Conjugate | MAB3402B, from mouse, biotin conjugate