MilliporeSigma
  • Discovery of Ketone-Based Covalent Inhibitors of Coronavirus 3CL Proteases for the Potential Therapeutic Treatment of COVID-19.
Discovery of Ketone-Based Covalent Inhibitors of Coronavirus 3CL Proteases for the Potential Therapeutic Treatment of COVID-19.
Journal of medicinal chemistry (2020-10-16)
Robert L Hoffman, Robert S Kania, Mary A Brothers, Jay F Davies, Rose A Ferre, Ketan S Gajiwala, Mingying He, Robert J Hogan, Kirk Kozminski, Lilian Y Li, Jonathan W Lockner, Jihong Lou, Michelle T Marra, Lennert J Mitchell, Brion W Murray, James A Nieman, Stephen Noell, Simon P Planken, Thomas Rowe, Kevin Ryan, George J Smith, James E Solowiej, Claire M Steppan, Barbara Taggart
ABSTRACT
The novel coronavirus disease COVID-19 that emerged in 2019 is caused by the virus SARS CoV-2 and named for its close genetic similarity to SARS CoV-1 that caused severe acute respiratory syndrome (SARS) in 2002. Both SARS coronavirus genomes encode two overlapping large polyproteins, which are cleaved at specific sites by a 3C-like cysteine protease (3CLpro) in a post-translational processing step that is critical for coronavirus replication. The 3CLpro sequences for CoV-1 and CoV-2 viruses are 100% identical in the catalytic domain that carries out protein cleavage. A research effort that focused on the discovery of reversible and irreversible ketone-based inhibitors of SARS CoV-1 3CLpro employing ligand-protease structures solved by X-ray crystallography led to the identification of 3 and 4. Preclinical experiments reveal 4 (PF-00835231) as a potent inhibitor of CoV-2 3CLpro with suitable pharmaceutical properties to warrant further development as an intravenous treatment for COVID-19.
Sigma-Aldrich
PF-00835231, ≥98% (HPLC)