MilliporeSigma
  • Platelet-Derived PDGFB Promotes Recruitment of Cancer-Associated Fibroblasts, Deposition of Extracellular Matrix and Tgfβ Signaling in the Tumor Microenvironment.
Platelet-Derived PDGFB Promotes Recruitment of Cancer-Associated Fibroblasts, Deposition of Extracellular Matrix and Tgfβ Signaling in the Tumor Microenvironment.
Cancers (2022-04-24)
Yanyu Zhang, Ehsan Manouchehri Doulabi, Melanie Herre, Jessica Cedervall, Qi Qiao, Zuoxiu Miao, Anahita Hamidi, Lars Hellman, Masood Kamali-Moghaddam, Anna-Karin Olsson
ABSTRACT
Platelets constitute a major reservoir of platelet-derived growth factor B (PDGFB) and are continuously activated in the tumor microenvironment, exposing tumors to the plethora of growth factors contained in platelet granules. To address the specific role of platelet-derived PDGFB in the tumor microenvironment, we have created a mouse model with conditional knockout of PDGFB in platelets (pl-PDGFB KO). Lack of PDGFB in platelets resulted in 10-fold lower PDGFB concentration in the tumor microenvironment, fewer cancer-associated fibroblasts and reduced deposition of the extracellular matrix (ECM) molecules fibronectin and collagen I in the orthotopic RIP1-Tag2 model for pancreatic neuroendocrine cancer. Myosin light chain phosphorylation, promoting cell contraction and, consequently, the mechano-induced release of active transforming growth factor (TGF) β from extracellular compartments, was reduced in tumors from pl-PDGFB KO mice. In agreement, TGFβ signaling, measured as phosphorylated Smad2, was significantly hampered in tumors from mice lacking PDGFB in their platelets, providing a plausible explanation for the reduced deposition of extracellular matrix. These findings indicate a major contribution of platelet-derived PDGFB to a malignant transformation of the tumor microenvironment and address for the first time the role of PDGFB released specifically from platelets in the remodeling of the ECM in tumors.
Sigma-Aldrich
Anti-PDGFB antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
IgG from goat serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder