MilliporeSigma
  • Preparation and Characterization of Polymer Composite Materials Based on PLA/TiO₂ for Antibacterial Packaging.
Preparation and Characterization of Polymer Composite Materials Based on PLA/TiO₂ for Antibacterial Packaging.
Polymers (2019-04-10)
Edwin A Segura González, Dania Olmos, Miguel Ángel Lorente, Itziar Vélaz, Javier González-Benito
ABSTRACT
Polymer composite materials based on polylactic acid (PLA) filled with titanium dioxide (TiO₂) nanoparticles were prepared. The aim of this work was to investigate the antibacterial action of TiO₂ against a strain of E. coli (DH5α) to obtain information on their potential uses in food and agro-alimentary industry. PLA/TiO₂ systems were prepared by a two-step process: Solvent casting followed by a hot-pressing step. Characterization was done as a function of particle size (21 nm and <100 nm) and particle content (0%, 1%, 5%, 10%, and 20%, wt %). Structural characterization carried out by X-ray diffraction (XRD) and Fourier Transformed Infrared spectroscopy (FTIR) did not reveal significant changes in polymer structure due to the presence of TiO₂ nanoparticles. Thermal characterization indicated that thermal transitions, measured by differential scanning calorimetry (DSC), did not vary, irrespective of size or content, whereas thermogravimetric analysis (TGA) revealed a slight increase in the temperature of degradation with particle content. Bacterial growth and biofilm formation on the surface of the composites against DH5α Escherichia coli was studied. Results suggested that the presence of TiO₂ nanoparticles decreases the amount of extracellular polymeric substance (EPS) and limits bacterial growth. The inhibition distances estimated with the Kirby-Bauer were doubled when 1% TiO₂ nanoparticles were introduced in PLA, though no significant differences were obtained for higher contents in TiO₂ NPs.
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis