MilliporeSigma
  • Phenethyl isothiocyanate reduces breast cancer stem cell-like properties by epigenetic reactivation of CDH1.
Phenethyl isothiocyanate reduces breast cancer stem cell-like properties by epigenetic reactivation of CDH1.
Oncology reports (2021-01-09)
Taolan Zhang, Weijuan Zhang, Meng Hao
ABSTRACT
Breast cancer is the most frequently diagnosed malignancy and leading cause of cancer-related deaths among women worldwide. Tumor recurrence, or metastasis, is caused by cancer stem cells and has a dismal prognosis for breast cancer patients. Thus, targeting breast cancer stem cells (BCSCs) for eradication is a potential method to improve clinical outcomes. Phenethyl isothiocyanate (PEITC) is a novel epigenetic regulator derived from cruciferous vegetables that has marked antitumor effects. However, the exact mechanism of these antitumor effects by PEITC is unknown. As breast cancer progresses, a tumor suppressor in the breast, cadherin 1 (CDH1), is silenced by hypermethylation of the promoter region, further promoting the stem cell-like properties of cancer. Herein, the ability of PEITC to reduce BCSC-like properties by epigenetic reactivation of CDH1 was investigated by multiple analyses such as MTT, colony formation and sphere formation assays, methylation-specific PCR, western blot analysis, Co-IP and qPCR. It was revealed that PEITC inhibited colony and mammosphere formation and decreased the expression of protein markers associated with BCSC-like properties via epigenetic reactivation of CDH1. Further exploration of this mechanism revealed inhibitory effects of PEITC on DNMTs and HDACs, which play a pivotal role in demethylating the hypermethylated CDH1 promoter region. Reactivated CDH1 suppressed the Wnt/β-catenin pathway which confers BCSC-properties in breast cancer cells. These findings suggest a novel method to eradicate BCSCs from breast cancer patients.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture