MilliporeSigma
  • WASp triggers mechanosensitive actin patches to facilitate immune cell migration in dense tissues.
WASp triggers mechanosensitive actin patches to facilitate immune cell migration in dense tissues.
Developmental cell (2021-12-18)
Florian Gaertner, Patricia Reis-Rodrigues, Ingrid de Vries, Miroslav Hons, Juan Aguilera, Michael Riedl, Alexander Leithner, Saren Tasciyan, Aglaja Kopf, Jack Merrin, Vanessa Zheden, Walter Anton Kaufmann, Robert Hauschild, Michael Sixt
ABSTRACT
When crawling through the body, leukocytes often traverse tissues that are densely packed with extracellular matrix and other cells, and this raises the question: How do leukocytes overcome compressive mechanical loads? Here, we show that the actin cortex of leukocytes is mechanoresponsive and that this responsiveness requires neither force sensing via the nucleus nor adhesive interactions with a substrate. Upon global compression of the cell body as well as local indentation of the plasma membrane, Wiskott-Aldrich syndrome protein (WASp) assembles into dot-like structures, providing activation platforms for Arp2/3 nucleated actin patches. These patches locally push against the external load, which can be obstructing collagen fibers or other cells, and thereby create space to facilitate forward locomotion. We show in vitro and in vivo that this WASp function is rate limiting for ameboid leukocyte migration in dense but not in loose environments and is required for trafficking through diverse tissues such as skin and lymph nodes.
Sigma-Aldrich
CK-666, ≥98% (HPLC), powder
Roche
DNase I, grade II, from bovine pancreas
Sigma-Aldrich
Arp2/3 Complex Inhibitor I, Inactive Control, CK-689, The Arp2/3 Complex Inhibitor I, Inactive Control, CK-689, also referenced under CAS 170930-46-8, controls the biological activity of Arp2/3. This small molecule/inhibitor is primarily used for Cell Structure applications.