MilliporeSigma
  • Trophoblast cell surface antigen 2 expression predicts outcome in oral squamous cell carcinomas.
Trophoblast cell surface antigen 2 expression predicts outcome in oral squamous cell carcinomas.
Oral diseases (2021-02-23)
Mauricio Rocha Dourado, Renato Assis Machado, Lívia Máris Ribeiro Paranaíba, Wilfredo Alejandro González-Arriagada, Sabrina Daniela da Silva, Íris Sawazaki-Calone, Edgard Graner, Tuula Salo, Ricardo D Coletta
ABSTRACT
Trophoblast cell surface antigen 2 (TROP2) has unclear clinical role in oral squamous cell carcinomas (OSCC). Here, we investigated the association of TROP2 immunoexpression with clinicopathological parameters and survival of OSCC patients. Cancer-specific survival (CSS) and disease-free survival (DFS) were assessed in a cohort composed of 266 OSCC. An independent cohort with 88 OSCC samples matched with the normal oral tissue, as well as 17 metastatic lymph nodes, was used for validation. Multivariate analysis showed TROP2 as an independent marker of favorable prognosis for both CSS (HR: 0.60, 95% CI: 0.40-0.90, p = .01) and DFS (HR: 0.57, 95% CI: 0.36-0.89, p = .01). Furthermore, TROP2 protein expression was significantly higher in morphologically normal tissues compared to primary tumors (p < .0001) and lymph node metastases (p = .001), and it was significantly associated with CSS (HR: 0.26, 95% CI: 0.09-0.74, p = .008) in the validation cohort. A pooled mRNA analysis performed on the Oncomine™ database confirmed the underexpression in OSCC compared with normal tissues (p = .014). In summary, our results point to a favorable prognostic significance of TROP2 overexpression in a large cohort of oral cancer patients, suggesting it as an attractive clinical marker.
Sigma-Aldrich
Anti-TACSTD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution