MilliporeSigma
  • Resolving macrophage polarization through distinct Ca2+ entry channel that maintains intracellular signaling and mitochondrial bioenergetics.
Resolving macrophage polarization through distinct Ca2+ entry channel that maintains intracellular signaling and mitochondrial bioenergetics.
iScience (2021-11-25)
Viviane Nascimento Da Conceicao, Yuyang Sun, Karthik Ramachandran, Arun Chauhan, Amritha Raveendran, Manigandan Venkatesan, Bony DeKumar, Soumya Maity, Neelanjan Vishnu, George A Kotsakis, Paul F Worley, Donald L Gill, Bibhuti B Mishra, Muniswamy Madesh, Brij B Singh
ABSTRACT
Transformation of naive macrophages into classically (M1) or alternatively (M2) activated macrophages regulates the inflammatory response. Here, we identified that distinct Ca2+ entry channels determine the IFNγ-induced M1 or IL-4-induced M2 transition. Naive or M2 macrophages exhibit a robust Ca2+ entry that was dependent on Orai1 channels, whereas the M1 phenotype showed a non-selective TRPC1 current. Blockade of Ca2+ entry suppresses pNF-κB/pJNK/STAT1 or STAT6 signaling events and consequently lowers cytokine production that is essential for M1 or M2 functions. Of importance, LPS stimulation shifted M2 cells from Orai1 toward TRPC1-mediated Ca2+ entry and TRPC1-/- mice exhibited transcriptional changes that suppress pro-inflammatory cytokines. In contrast, Orai1-/- macrophages showed a decrease in anti-inflammatory cytokines and exhibited a suppression of mitochondrial oxygen consumption rate and inhibited mitochondrial shape transition specifically in the M2 cells. Finally, alterations in TRPC1 or Orai1 expression determine macrophage polarization suggesting a distinct role of Ca2+ channels in modulating macrophage transformation.
Product Description
Roche
Cell Proliferation Kit I (MTT)
Sigma-Aldrich
Anti-TRPC3 antibody produced in rabbit, affinity isolated antibody, lyophilized powder