MilliporeSigma
  • Early Prediction of Graft Outcomes After Kidney Transplantation From Donors After Circulatory Death: Biomarkers and Transplantation Characteristics.
Early Prediction of Graft Outcomes After Kidney Transplantation From Donors After Circulatory Death: Biomarkers and Transplantation Characteristics.
Transplantation proceedings (2019-11-17)
Anne-Sophie Truche, Candice Trocme, Sabrina Vergnaud, Bénédicte Janbon, Diane Giovannini, Paolo Malvezzi, Xavier Moreau-Gaudry, Lionel Rostaing, Rachel Tetaz
ABSTRACT
This study aimed to identify transplantation characteristics and biomarkers that predict outcomes for kidney transplant (KT) patients from donors after circulatory death (DCDs). Consecutive patients receiving a KT from a DCD in our center between 2014 and 2016 were included; the reference population was recipients with a living donor KT. The urinary tubular injury biomarker-to-creatinine ratio and serum lactate dehydrogenase (LDH) were measured at post-transplant days 1 and 3. The primary outcome was the occurrence of delayed graft function (DGF). Descriptive and receiver operating characteristic analyses were performed. Forty-one patients were included in the analysis: 15 (36.59%) DCD KTs (9 of which suffered from DGF) and 26 (63.41%) living donor KTs. For the primary endpoint, neutrophil gelatinase-associated lipocalin, N-acetyl-beta-D-glucosaminidase, urinary tubular injury biomarker-to-creatinine ratio, and LDH areas under the curve were 1 and 0.96 (95% confidence interval: 0.84-1.0), 1 and 0.92 (95% confidence interval: 0.73-1.0), respectively. Among the transplant characteristics, only the 30-minute resistive index on the perfusion machine was significantly higher in DCD KTs with DGF vs those without DGF (0.26 mm Hg/mL/min [0.20; 0.32] vs 0.14 mm Hg/mL/min [0.12; 0.16], P = .05). Median 3-month creatinine clearance among DGF DCD KTs was 49 mL/min/1.73 m2 [IQR: 42; 65] and 65 mL/min/1.73 m2 [IQR: 62; 66] among DCD KTs without DGF (P = .22). In the DCD KT population, clinical and biological markers were identified that provided predictive tools for DGF. Thus, systematic measurement of these biomarkers, particularly LDH, could improve the management of kidney graft recipients' immunosuppressive therapy.
Product Description
Roche
N-Acetyl-β-D-Glucosaminidase (NAG), sufficient for ~50 tests