MilliporeSigma
MCC950 closes the active conformation of NLRP3 to an inactive state.
Nature chemical biology (2019-05-16)
Ana Tapia-Abellán, Diego Angosto-Bazarra, Helios Martínez-Banaclocha, Carlos de Torre-Minguela, Jose P Cerón-Carrasco, Horacio Pérez-Sánchez, Juan I Arostegui, Pablo Pelegrin
ABSTRACT
NLRP3 (NOD-like receptor pyrin domain-containing protein 3) is an innate immune sensor that contributes to the development of different diseases, including monogenic autoinflammatory syndromes, gout, atherosclerosis, and Alzheimer's disease. The molecule sulfonylurea MCC950 is a NLRP3 inflammasome inhibitor with potential clinical utility. However, the mechanism of action of MCC950 remains unknown. Here, we characterize the mechanism of action of MCC950 in both wild-type and autoinflammatory-related NLRP3 mutants, and demonstrate that MCC950 closes the 'open' conformation of active NLRP3.
Sigma-Aldrich
Protein A, Peroxidase Conjugate, This Protein A, Peroxidase Conjugate is validated for use in Direct Immunoassay.