MilliporeSigma
  • Kinetochore life histories reveal an Aurora-B-dependent error correction mechanism in anaphase.
Kinetochore life histories reveal an Aurora-B-dependent error correction mechanism in anaphase.
Developmental cell (2021-11-11)
Onur Sen, Jonathan U Harrison, Nigel J Burroughs, Andrew D McAinsh
ABSTRACT
Chromosome mis-segregation during mitosis leads to aneuploidy, which is a hallmark of cancer and linked to cancer genome evolution. Errors can manifest as "lagging chromosomes" in anaphase, although their mechanistic origins and likelihood of correction are incompletely understood. Here, we combine lattice light-sheet microscopy, endogenous protein labeling, and computational analysis to define the life history of >104 kinetochores. By defining the "laziness" of kinetochores in anaphase, we reveal that chromosomes are at a considerable risk of mis-segregation. We show that the majority of lazy kinetochores are corrected rapidly in anaphase by Aurora B; if uncorrected, they result in a higher rate of micronuclei formation. Quantitative analyses of the kinetochore life histories reveal a dynamic signature of metaphase kinetochore oscillations that forecasts their anaphase fate. We propose that in diploid human cells chromosome segregation is fundamentally error prone, with an additional layer of anaphase error correction required for stable karyotype propagation.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
MKLP-2 Inhibitor, Paprotrain, The MKLP-2 Inhibitor, Paprotrain controls the biological activity of MKLP-2. This small molecule/inhibitor is primarily used for Cell Signaling applications.
Sigma-Aldrich
Aurora Kinase Inhibitor VI, ZM447439, The Aurora Kinase Inhibitor VI, ZM447439, also referenced under CAS 331771-20-1, controls the biological activity of Aurora Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.