MilliporeSigma
  • Interferon-gamma limits Th1 lymphocyte adhesion to inflamed endothelium: a nitric oxide regulatory feedback mechanism.
Interferon-gamma limits Th1 lymphocyte adhesion to inflamed endothelium: a nitric oxide regulatory feedback mechanism.
European journal of immunology (2008-04-17)
M Ursula Norman, Lori Zbytnuik, Paul Kubes
ABSTRACT
CD4(+) T helper (Th1 and Th2) cell localization to a site of inflammation is important for the development, maintenance and regulation of an immune response. The factors that regulate Th1 and Th2 cell recruitment into tissue are not fully understood. The aim of the present study was to examine the effect of different cytokine microenvironments on the recruitment of Th1 and Th2 lymphocytes into tissue. Fluorescently labelled Th1 or Th2 lymphocyte-endothelial interactions were observed via intravital microscopy of the cytokine-treated cremaster muscle. Our results show that TNF-alpha alone is sufficient to maximally recruit Th1 cells. Surprisingly, treatment with TNF-alpha + IFN-gamma significantly decreased Th1 adhesion and emigration in comparison to TNF-alpha treatment alone. The decreased adhesion of Th1 cells in response to TNF-alpha + IFN-gamma reflected a decreased ability to bind to ICAM-1 and was iNOS-dependent. This phenomenon was not observed with Th2 cells. These results suggest that IFN-gamma may play a key immunomodulatory role in the recruitment of different T lymphocyte subsets. Indeed, blockade of IFN-gamma or iNOS function during the Th1-mediated contact hypersensitivity response resulted in an acceleration and exacerbation of the late-phase inflammatory response.
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone PS/2, clone PS/2, Chemicon®, from rat