MilliporeSigma
  • Identification of a conserved region of Plasmodium falciparum MSP3 targeted by biologically active antibodies to improve vaccine design.
Identification of a conserved region of Plasmodium falciparum MSP3 targeted by biologically active antibodies to improve vaccine design.
The Journal of infectious diseases (2004-08-06)
Subhash Singh, Soe Soe, Juan-Pedro Mejia, Christian Roussilhon, Michael Theisen, Giampietro Corradin, Pierre Druilhe
ABSTRACT
Merozoite surface protein 3 (MSP3) is a target of antibody-dependent cellular inhibition (ADCI), a protective mechanism against Plasmodium falciparum malaria. From the C-terminal half of the molecule, 6 overlapping peptides were chosen to characterize human immune responses. Each peptide defined at least 1 non-cross-reactive B cell epitope. Distinct patterns of antibody responses, by level and IgG subclass distribution, were observed in inhabitants of a malaria-endemic area. Antibodies affinity purified toward each peptide differed in their functional capacity to mediate parasite killing in ADCI assays: 3 of 6 overlapping peptides had a major inhibitory effect on parasite growth. This result was confirmed by the passive transfer of anti-MSP3 antibodies in vivo in a P. falciparum mouse model. T helper cell epitopes were identified in each peptide. Antigenicity and functional assays identified a 70-amino acid conserved domain of MSP3 as a target of biologically active antibodies to be included in future vaccine constructs based on MSP3.
Sigma-Aldrich
Mouse Anti-Human IgG2 Antibody, clone HP6002, Fc, clone HP6002, Chemicon®, from mouse