MilliporeSigma
  • Attenuated succinate accumulation relieves neuronal injury induced by hypoxia in neonatal mice.
Attenuated succinate accumulation relieves neuronal injury induced by hypoxia in neonatal mice.
Cell death discovery (2022-03-31)
Mengdi Zhang, Yao Cheng, Yujie Zhai, Yaru Cui, Wenshen Zhang, Hongwei Sun, Wenyu Xin, Ling Zhou, Xue Gao, Shucui Li, Hongliu Sun
ABSTRACT
Hypoxia causes neonatal neuronal damage. However, the underlying mechanism remains unclear. This study aimed to explore the changes in succinate levels and identify the mechanisms underlying their contribution to hypoxia-induced damage in newborn mice. The neonatal C57BL/6J mouse hypoxia model was used in our study. We evaluated the levels of succinate, iron, reactive oxygen species (ROS), and mitochondrial ROS, and assessed mitophagy, neuronal damage, and learning and memory function, after hypoxia treatment. The neonatal mice showed increased succinate levels in the early hypoxia stage, followed by increased levels of oxidative stress, iron stress, neuronal damage, and cognitive deficits. Succinate levels were significantly reduced following treatment with inhibitors of succinate dehydrogenase (SDH), purine nucleotide cycle (PNC), and malate/aspartate shuttle (MAS), with the corresponding attenuation of oxidative stress, iron stress, neuronal damage, and cognitive impairment. Reversal catalysis of SDH through fumarate from the PNC and MAS pathways might be involved in hypoxia-induced succinate accumulation. Succinate accumulation in the early period after hypoxia may crucially contribute to oxidative and iron stress. Relieving succinate accumulation at the early hypoxia stage could prevent neuronal damage and cognitive impairment in neonatal hypoxia.