MilliporeSigma
  • A conserved strategy for inducing appendage regeneration in moon jellyfish, Drosophila, and mice.
A conserved strategy for inducing appendage regeneration in moon jellyfish, Drosophila, and mice.
eLife (2021-12-08)
Michael J Abrams, Fayth Hui Tan, Yutian Li, Ty Basinger, Martin L Heithe, Anish Sarma, Iris T Lee, Zevin J Condiotte, Misha Raffiee, John O Dabiri, David A Gold, Lea Goentoro
ABSTRACT
Can limb regeneration be induced? Few have pursued this question, and an evolutionarily conserved strategy has yet to emerge. This study reports a strategy for inducing regenerative response in appendages, which works across three species that span the animal phylogeny. In Cnidaria, the frequency of appendage regeneration in the moon jellyfish Aurelia was increased by feeding with the amino acid L-leucine and the growth hormone insulin. In insects, the same strategy induced tibia regeneration in adult Drosophila. Finally, in mammals, L-leucine and sucrose administration induced digit regeneration in adult mice, including dramatically from mid-phalangeal amputation. The conserved effect of L-leucine and insulin/sugar suggests a key role for energetic parameters in regeneration induction. The simplicity by which nutrient supplementation can induce appendage regeneration provides a testable hypothesis across animals.
Sigma-Aldrich
Insulin human, meets USP testing specifications
Sigma-Aldrich
Anti-alpha-Tubulin Antibody, tyrosinated, clone YL1/2, clone YL 1/2, from rat