MilliporeSigma
  • Response prediction in patients with gastric and esophagogastric adenocarcinoma under neoadjuvant chemotherapy using targeted gene expression analysis and next-generation sequencing in pre-therapeutic biopsies.
Response prediction in patients with gastric and esophagogastric adenocarcinoma under neoadjuvant chemotherapy using targeted gene expression analysis and next-generation sequencing in pre-therapeutic biopsies.
Journal of cancer research and clinical oncology (2022-03-06)
Karsten Kleo, Vladimir M Jovanovic, Alexander Arndold, Annika Lehmann, Hedwig Lammert, Erika Berg, Hannah Harloff, Christoph Treese, Michael Hummel, Severin Daum
ABSTRACT
Perioperative chemo-(radio-) therapy is the accepted standard in European patients with locally advanced adenocarcinoma of the esophagogastric junction or stomach (AEG/AS). However, 30-85% of patients do not respond to this treatment. The aim of our study was the identification of predictive biomarkers in pre-therapeutic endoscopic tumor biopsies from patients with histopathologic response (Becker-1) versus non-response (Becker-2/3) to preoperative chemotherapy. Formalin-fixed paraffin-embedded biopsies from 36 Caucasian patients (Becker-1 n = 11, Becker-2 n = 7, Becker-3 n = 18) with AEG/AS, taken prior to neoadjuvant chemotherapy were selected. For RNA expression analysis, we employed the NanoString nCounter System. To identify genomic alterations like single nucleotide variants (SNV), copy number variation (CNV) and fusion events, we used Illumina TST170 gene panel. For HER2 and FGFR2 protein expression, immunostaining was performed. Furthermore, we analyzed the microsatellite instability (MSI) and Epstein-Barr virus (EBV) infection status by EBER in situ hybridization. Heat map and principal component analyses showed no clustering by means of gene expression according to regression grade. Concerning two recently proposed predictive markers, our data showed equal distribution for MSI (Becker-1: 2; Becker-2: 1; Becker-3: 3; out of 29 tested) and EBV infection was rare (1/32). We could not reveal discriminating target genes concerning SNV, but found a higher mutational burden in non-responders versus responders and fusion (in 6/14) and CNV events (in 5/14) exclusively in Becker-3. Although we could not identify discriminating target genes, our data suggest that molecular alterations are in general more prevalent in patients with AEG/AS belonging to the non-responding Becker group 3.
Sigma-Aldrich
Anti-FGFR2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution