MilliporeSigma
  • An Early Disturbance in Serotonergic Neurotransmission Contributes to the Onset of Parkinsonian Phenotypes in Drosophila melanogaster.
An Early Disturbance in Serotonergic Neurotransmission Contributes to the Onset of Parkinsonian Phenotypes in Drosophila melanogaster.
Cells (2022-05-15)
Rafaella V Zárate, Sergio Hidalgo, Nicole Navarro, Daniela Molina-Mateo, Duxan Arancibia, Francisca Rojo-Cortés, Carlos Oliva, María Estela Andrés, Pedro Zamorano, Jorge M Campusano
ABSTRACT
Parkinson's disease (PD) is a neurodegenerative disease characterized by motor symptoms and dopaminergic cell loss. A pre-symptomatic phase characterized by non-motor symptoms precedes the onset of motor alterations. Two recent PET studies in human carriers of mutations associated with familial PD demonstrate an early serotonergic commitment-alteration in SERT binding-before any dopaminergic or motor dysfunction, that is, at putative PD pre-symptomatic stages. These findings support the hypothesis that early alterations in the serotonergic system could contribute to the progression of PD, an idea difficult to be tested in humans. Here, we study some components of the serotonergic system during the pre-symptomatic phase in a well-characterized Drosophila PD model, Pink1B9 mutant flies. We detected lower brain serotonin content in Pink1B9 flies, accompanied by reduced activity of SERT before the onset of motor dysfunctions. We also explored the consequences of a brief early manipulation of the serotonergic system in the development of motor symptoms later in aged animals. Feeding young Pink1B9 flies with fluoxetine, a SERT blocker, prevents the loss of dopaminergic neurons and ameliorates motor impairment observed in aged mutant flies. Surprisingly, the same pharmacological manipulation in young control flies results in aged animals exhibiting a PD-like phenotype. Our findings support that an early dysfunction in the serotonergic system precedes and contributes to the onset of the Parkinsonian phenotype in Drosophila.
Sigma-Aldrich
Anti-Tryptophan Hydroxylase Antibody, Chemicon®, from sheep
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit