MilliporeSigma
  • Targeted intestinal deletion of Rho guanine nucleotide exchange factor 7, βPIX, impairs enterocyte proliferation, villus maturation, and mucosal defenses in mice.
Targeted intestinal deletion of Rho guanine nucleotide exchange factor 7, βPIX, impairs enterocyte proliferation, villus maturation, and mucosal defenses in mice.
American journal of physiology. Gastrointestinal and liver physiology (2021-02-11)
Kunrong Cheng, Shannon M Larabee, Mazen Tolaymat, Marie Hanscom, Aaron C Shang, Alyssa Schledwitz, Shien Hu, Cinthia B Drachenberg, Min Zhan, Ahmed Chahdi, Jean-Pierre Raufman
ABSTRACT
Rho guanine nucleotide exchange factors (RhoGEFs) regulate Rho GTPase activity and cytoskeletal and cell adhesion dynamics. βPix, a CDC42/RAC family RhoGEF encoded by ARHGEF7, is reported to modulate human colon cancer cell proliferation and postwounding restitution of rat intestinal epithelial monolayers. We hypothesized that βPix plays a role in maintaining intestinal epithelial homeostasis. To test this hypothesis, we examined βPix distribution in the human and murine intestine and created mice with intestinal epithelial-selective βPix deletion [βPixflox/flox/Tg(villin-Cre); Arhgef7 CKO mice]. Using Arhgef7 conditional knockout (CKO) and control mice, we investigated the consequences of βPix deficiency in vivo on intestinal epithelial and enteroid development, dextran sodium sulfate-induced mucosal injury, and gut permeability. In normal human and murine intestines, we observed diffuse cytoplasmic and moderate nuclear βPix immunostaining in enterocytes. Arhgef7 CKO mice were viable and fertile, with normal gross intestinal architecture but reduced small intestinal villus height, villus-to-crypt ratio, and goblet cells; small intestinal crypt cells had reduced Ki67 staining, compatible with impaired cell proliferation. Enteroids derived from control mouse small intestine were viable for more than 20 passages, but those from Arhgef7 CKO mice did not survive beyond 24 h despite addition of Wnt proteins or conditioned media from normal enteroids. Adding a Rho kinase (ROCK) inhibitor partially rescued CKO enteroid development. Compared with littermate control mice, dextran sodium sulfate-treated βPix-deficient mice lost more weight and had greater impairment of intestinal barrier function, and more severe colonic mucosal injury. These findings reveal βPix expression is important for enterocyte development, intestinal homeostasis, and resistance to toxic injury.NEW & NOTEWORTHY To explore the role of βPix, a guanine nucleotide exchange factor encoded by ARHGEF7, in intestinal development and physiology, we created mice with intestinal epithelial cell Arhgef7/βPix deficiency. We found βPix essential for normal small intestinal epithelial cell proliferation, villus development, and mucosal resistance to injury. Moreover, Rho kinase signaling mediated developmental arrest observed in enteroids derived from βPix-deficient small intestinal crypts. Our studies provide insights into the role Arhgef7/βPix plays in intestinal epithelial homeostasis.
Sigma-Aldrich
Anti-ARHGEF7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Beta Pix, SH3 Domain Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Active-β-Catenin Antibody, clone 8E7, Alexa Fluor 555, clone 8E7, from mouse, ALEXA FLUOR 555