MilliporeSigma
  • Direct Comparison of Poly(ethylene glycol) and Phosphorylcholine Drug-Loaded Nanoparticles In Vitro and In Vivo.
Direct Comparison of Poly(ethylene glycol) and Phosphorylcholine Drug-Loaded Nanoparticles In Vitro and In Vivo.
Biomacromolecules (2020-04-29)
Janina-Miriam Noy, Fan Chen, Dewan T Akhter, Zachary H Houston, Nicholas L Fletcher, Kristofer J Thurecht, Martina H Stenzel
ABSTRACT
Phosphorylcholine is known to repel the absorption of proteins onto surfaces, which can prevent the formation of a protein corona on the surface of nanoparticles. This can influence the fate of nanoparticles used for drug delivery. This material could therefore serve as an alternative to poly(ethylene glycol) (PEG). Herein, the synthesis of different particles prepared by polymerization-induced self-assembly (PISA) coated with either poly(ethylene glycol) (PEG) or zwitterionic 2-methacryloyloxyethyl phosphorylcholine (MPC) and 4-(N-(S-penicillaminylacetyl)amino) phenylarsenonous acid (PENAO) was reported. The anticancer drug 4-(N-(S-penicillaminylacetyl)amino) phenylarsenonous acid (PENAO) was conjugated to the shell-forming block. Interactions of the different coated nanoparticles, which present comparable sizes and size distributions (76-85 nm, PDI = 0.067-0.094), with two-dimensional (2D) and three-dimensional (3D) cultured cells were studied, and their cytotoxicities, cellular uptakes, spheroid penetration, and cell localization profiles were analyzed. While only a minimal difference in behaviour was observed for nanoparticles assessed using in vitro experiment (with PEG-co- PENAO-coated micelles showing slightly higher cytotoxicity and better spheroid penetration and cell localization ability), the effect of the different physicochemical properties between nanoparticles had a more dramatic effect on in vivo biodistribution. After 1 h of injection, the majority of the MPC-co-PENAO-coated nanoparticles were found to accumulate in the liver, making this particle system unfeasible for future biological studies.
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 35,000
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 20,000
Sigma-Aldrich
Poly(ethylene glycol), tested according to Ph. Eur., 400