MilliporeSigma
  • An Insight into Sesamolin: Physicochemical Properties, Pharmacological Activities, and Future Research Prospects.
An Insight into Sesamolin: Physicochemical Properties, Pharmacological Activities, and Future Research Prospects.
Molecules (Basel, Switzerland) (2021-10-14)
Reny Rosalina, Natthida Weerapreeyakul
ABSTRACT
Sesame seeds are rich in lignan content and have been well-known for their health benefits. Unlike the other sesame lignan compounds (i.e., sesamin and sesamol), the study of the pharmacological activity of sesamolin has not been explored widely. This review, therefore, summarizes the information related to sesamolin's pharmacological activities, and the mechanism of action. Moreover, the influence of its physicochemical properties on pharmacological activity is also discussed. Sesamolin possessed neuroprotective activity against hypoxia-induced reactive oxygen species (ROS) and oxidative stress in neuron cells by reducing the ROS and inhibiting apoptosis. In skin cancer, sesamolin exhibited antimelanogenesis by affecting the expression of the melanogenic enzymes. The anticancer activity of sesamolin based on antiproliferation and inhibition of migration was demonstrated in human colon cancer cells. In addition, treatment with sesamolin could stimulate immune cells to enhance the cytolytic activity to kill Burkitt's lymphoma cells. However, the toxicity and safety of sesamolin have not been reported. And there is also less information on the experimental study in vivo. The limited aqueous solubility of sesamolin becomes the main problem, which affects its pharmacological activity in the in vitro experiment and clinical efficacy. Therefore, solubility enhancement is needed for further investigation and determination of its pharmacological activity profiles. Since there are fewer reports studying this issue, it could become a future prospective research opportunity.
Sigma-Aldrich
Sesamolin, ≥97%, from Sesamum indicum (sesame)