MilliporeSigma
  • Pharmacological characterization of AC-262536, a novel selective androgen receptor modulator.
Pharmacological characterization of AC-262536, a novel selective androgen receptor modulator.
The Journal of steroid biochemistry and molecular biology (2008-01-01)
Fabrice Piu, Luis R Gardell, Thomas Son, Nathalie Schlienger, Birgitte W Lund, Hans H Schiffer, Kim E Vanover, Robert E Davis, Roger Olsson, Stefania Risso Bradley
ABSTRACT
Because of the limitations and liabilities of current testosterone therapies, non-steroidal tissue-selective androgen receptor modulators may provide a clinically meaningful advance in therapy. Using a functional cell-based assay AC-262536 was identified as a potent and selective AR ligand, with partial agonist activity relative to the natural androgen testosterone. A 2-week chronic study in castrated male rats indicated that AC-262536 significantly improves anabolic parameters in these animals, especially in stimulating the growth of the levator ani and in suppressing elevated LH levels. In sharp contrast to testosterone, AC-262536 has weak androgenic effects, as measured by prostate and seminal vesicle weights. Thus, AC-262536 represents a novel class of selective androgen receptor modulators (SARMs) with beneficial anabolic effects.
Sigma-Aldrich
AC-262536, ≥95% (HPLC)