MilliporeSigma
Polyethylene glycol as a cause of anaphylaxis.
Allergy, asthma, and clinical immunology : official journal of the Canadian Society of Allergy and Clinical Immunology (2016-12-22)
Katharina Wylon, Sabine Dölle, Margitta Worm
ABSTRACT
Polyethylene glycols (PEGs) or macrogols are polyether compounds and are widely used as additives in pharmaceuticals, cosmetics, and food. We report on a Caucasian patient experiencing recurrent severe allergic reactions to several drugs. An extensive diagnostic workup including skin prick tests, intradermal tests (IDT) and a double-blind oral challenge was performed to identify the trigger of anaphylaxis. In the present case hypersensitivity to the additive polyethylene glycol was confirmed by an IDT suggesting an Immunoglobulin E-dependent mechanism as a cause of the reaction. Potential life-threatening hypersensitivity reactions to hidden molecules like macrogol may be underdiagnosed. Cases of immediate-type PEG hypersensitivity were reported with increasing frequency. The awareness regarding the allergenic potential of PEG should be raised and a proper product labelling is crucial to prevent PEG mediated hypersensitivity.
Sigma-Aldrich
Poly(ethylene glycol), tested according to Ph. Eur., 400
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 20,000
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 35,000