MilliporeSigma
  • Oxidation reactions of acetylenic fatty esters with selenium dioxide/tert-butyl hydroperoxide.
Oxidation reactions of acetylenic fatty esters with selenium dioxide/tert-butyl hydroperoxide.
Lipids (1997-11-14)
M S Jie, M K Pasha, M S Alam
ABSTRACT
Reaction of methyl undec-10-ynoate (1) with selenium dioxide/tert-butyl hydroperoxide (TBHP) in aqueous dioxane gave methyl 9-oxo-undec-10-ynoate (2, 9%) and 9-hydroxy-undec-10-ynoate (3, 60%), while methyl octadec-9-ynoate (4) yielded mixtures of positional isomers of mono-keto (viz. methyl 8-oxo- and 11-oxo-octadec-9-ynoate, 5, 5%), hydroxy-keto (viz. methyl 8-hydroxy-11-oxo- and 11-hydroxy-8-oxo-octadec-9-ynoate, 6, 10%), and dihydroxy (viz. methyl 8,11-dihydroxy-octadec-9-ynoate, 7, 24%) derivatives. Similar treatment of a conjugated diacetylenic fatty ester (methyl octadeca-6,8-diynoate, 8) furnished a mixture of methyl 5-oxo- and 10-oxo-octadeca-6,8-diynoate (9, 12%) and a complex mixture of very polar products. Reaction of methyl octadec-11E-en-9-ynoate (methyl santalbate) (10) with selenium dioxide/TBHP in aqueous dioxane gave exclusively a mixture of regiospecific products, viz. methyl 8-oxo-octadec-11(E)Z-en-9-ynoate (11, 6%) and methyl 8-hydroxy-octadec-11E-en-9-ynoate (12, 70%). The structures of the various products were determined by a combination of spectroscopic and mass spectral analyses.
Sigma-Aldrich
Selenium dioxide, (sublimed) for synthesis