MilliporeSigma
  • Transient Agarose Spot (TAS) Assay: A New Method to Investigate Cell Migration.
Transient Agarose Spot (TAS) Assay: A New Method to Investigate Cell Migration.
International journal of molecular sciences (2022-02-27)
Apor Veres-Székely, Domonkos Pap, Beáta Szebeni, László Őrfi, Csenge Szász, Csenge Pajtók, Eszter Lévai, Attila J Szabó, Ádám Vannay
ABSTRACT
Fibroblasts play a central role in diseases associated with excessive deposition of extracellular matrix (ECM), including idiopathic pulmonary fibrosis. Investigation of different properties of fibroblasts, such as migration, proliferation, and collagen-rich ECM production is unavoidable both in basic research and in the development of antifibrotic drugs. In the present study we developed a cost-effective, 96-well plate-based method to examine the migration of fibroblasts, as an alternative approach to the gold standard scratch assay, which has numerous limitations. This article presents a detailed description of our transient agarose spot (TAS) assay, with instructions for its routine application. Advantages of combined use of different functional assays for fibroblast activation in drug development are also discussed by examining the effect of nintedanib-an FDA approved drug against IPF-on lung fibroblasts.
Sigma-Aldrich
Diaphorase from Clostridium kluyveri, recombinant, expressed in E. coli, Animal-component free