MilliporeSigma
  • Melatonin added to freezing diluent improves canine (Bulldog) sperm cryosurvival.
Melatonin added to freezing diluent improves canine (Bulldog) sperm cryosurvival.
Reproduction & fertility (2020-11-04)
Julio A Martinez-Rodriguez, Francisco J Carbajal, Rocio Martinez-De-Anda, Alicia Alcantar-Rodriguez, Alfredo Medrano
ABSTRACT
Cryopreservation compromises the capacity of sperm fertilizing due to a series of alterations in the structure and physiology of the sperm. The use of antioxidants, such as melatonin, added to freezing media, may help to reduce sperm cryoinjury. To test the effect of melatonin on Bulldog (Canis lupus familiaris) sperm cryosurvival, spermatozoa were diluted in a standard freezing medium and cooled to 5°C. Then, more freezing medium was added to obtain 200 × 106 cells/mL, and 5% glycerol. Diluted spermatozoa were treated with melatonin (0.0, 0.0005, 0.002, and 0.0035 mol/L), and packaged in 0.25 mL straws, which were further cooled to -5°C before freezing in liquid nitrogen. Thawing was carried out at 70°C for 5 s, and the progressive motility, viability, plasma membrane integrity, acrosome integrity, capacitation status, and plasma membrane fluidity of the spermatozoa (at 37°C) were assessed. Data were analyzed using ANOVA to detect differences between the melatonin doses. There were statistical differences (P < 0.05) in the percentage of sperm having hyper-fluid membranes, intact acrosome, capacitated acrosome-intact, and acrosome-reacted. The values for the high melatonin doses (0.002 and 0.0035 mol/L) were better than for the low melatonin doses (0.0 and 0.0005 mol/L). In conclusion, 0.002 and 0.0035 mol/L of melatonin improved the cryosurvival of sperm from male bulldogs. Preservation of sperm by freezing enables breeding of individuals geographically separated; protocols for the dog may be used to preserve the semen from threatened wild canids. To improve fertility of female dogs that become pregnant with frozen and then defrosted sperm, these cells must survive that process which can be damaging whilst keeping their ability to fertilize. Antioxidants are substances capable of retarding or preventing the oxidation of any oxidizing substrate such as lipids, proteins, and DNA, which are structural compounds of the sperm. The use of antioxidants, added to freezing media, may provide the sperm the capacity to neutralize oxidative compounds, such as reactive oxygen species, produced during the freezing and thawing process. In this work we tested different levels of melatonin, a natural antioxidant, on dog (English Bulldog) sperm survival and quality after freezing. We found that adding melatonin to the freezing media improved sperm quality after thawing.
Sigma-Aldrich
Lectin from Pisum sativum (pea), FITC conjugate, lyophilized powder