MilliporeSigma
  • Spectroscopic analysis of keratin endogenous signal for skin multiphoton microscopy: erratum.
Spectroscopic analysis of keratin endogenous signal for skin multiphoton microscopy: erratum.
Optics express (2005-08-22)
A Pena, M Strupler, T Boulesteix, G Godeau, Marie-Claire Schanne-Klein
ABSTRACT
We present corrected versions of the list of authors and of Section 2.1.
Sigma-Aldrich
Keratin from human epidermis, aqueous solution
Pricing and availability is not currently available.