MilliporeSigma
  • Analysis of differential gene expression patterns in colon cancer and cancer stroma using microdissected tissues.
Analysis of differential gene expression patterns in colon cancer and cancer stroma using microdissected tissues.
Gastroenterology (2005-02-03)
Yuko Sugiyama, Buckminster Farrow, Carlos Murillo, Jing Li, Hiroaki Watanabe, Kazuo Sugiyama, B Mark Evers
ABSTRACT
The surrounding cancer stroma is increasingly recognized as playing an important role in cancer proliferation, invasion, and metastasis. Here, we analyzed patterns of gene expression in colon cancer cells, surrounding stroma, and normal mucosa and normal stroma using laser capture microdissection. Tissues were fixed from 11 patients undergoing colorectal resection for cancer, and laser capture microdissection was performed. Samples (4 per patient) were extracted for RNA, which was then used for focused gene arrays. In addition, protein expression of selected genes was determined by Western blot or immunohistochemistry. We showed differential expression patterns in cancer cells and surrounding stroma compared with their normal counterparts. Genes contributing to angiogenesis, cell cycle regulation, and proliferation were significantly increased in cancer cells compared with the adjacent normal mucosa. Genes contributing to angiogenesis, cancer invasion, and metastasis were significantly increased in surrounding cancer stromal cells compared with normal stroma. Delineating the differential patterns of gene expression between colon cancers and the surrounding reactive stroma will better determine the role of the stromal components in the progression of colon cancers and may lead to chemopreventive therapy targeted specifically to the stroma.
Sigma-Aldrich
Anti-TIMP-1 Antibody, clone 102D1, clone 102D1, Chemicon®, from mouse