MilliporeSigma
  • Peripheral or central administration of motilin suppresses LH release in female rats: a novel role for motilin.
Peripheral or central administration of motilin suppresses LH release in female rats: a novel role for motilin.
Journal of neuroendocrinology (2000-05-03)
H Tsukamura, S Tsukahara, F Maekawa, R Moriyama, B A Reyes, T Sakai, Y Niwa, D L Foster
ABSTRACT
Motilin is secreted in a clear episodic pattern during fasting or during the interdigestive phase, but feeding promptly stops this secretory pattern, and plasma concentrations of motilin decrease. We have previously determined that fasting markedly suppresses pulsatile luteinizing hormone (LH) secretion in female rats in the presence of oestrogen. In the present study, we wished to learn if motilin may mediate the fasting-induced suppression of LH secretion by determining the effects of motilin administration on LH release and on food intake. Intravenous (i.v.) injection of motilin (37 nmol/rat) suppressed LH release and significantly decreased mean LH concentrations both in ovariectomized (OVX) and oestradiol-implanted ovariectomized (OVX+E2) rats. Food intake was significantly increased by i.v. motilin injection in OVX rats, but not in OVX+E2 rats. It is likely that motilin inhibits LH release via inhibition of the gonadotrophin-releasing hormone (GnRH)-releasing mechanism at the hypothalamic level, because motilin (3.7 nmol/rat) also suppressed LH secretion when centrally administered, and because LH release in i.v. motilin-treated rats increased in response to exogenous GnRH. These results suggest that motilin may be a peripheral signal for the suppression of LH secretion through central sensors.
Sigma-Aldrich
Motilin porcine, ≥97% (HPLC)