MilliporeSigma
  • Tight function zonula occludens-3 regulates cyclin D1-dependent cell proliferation.
Tight function zonula occludens-3 regulates cyclin D1-dependent cell proliferation.
Molecular biology of the cell (2011-03-18)
Christopher T Capaldo, Stefan Koch, Michael Kwon, Oskar Laur, Charles A Parkos, Asma Nusrat
ABSTRACT
Coordinated regulation of cell proliferation is vital for epithelial tissue homeostasis, and uncontrolled proliferation is a hallmark of carcinogenesis. A growing body of evidence indicates that epithelial tight junctions (TJs) play a role in these processes, although the mechanisms involved are poorly understood. In this study, we identify and characterize a novel plasma membrane pool of cyclin D1 with cell-cycle regulatory functions. We have determined that the zonula occludens (ZO) family of TJ plaque proteins sequesters cyclin D1 at TJs during mitosis, through an evolutionarily conserved class II PSD-95, Dlg, and ZO-1 (PDZ)-binding motif within cyclin D1. Disruption of the cyclin D1/ZO complex through mutagenesis or siRNA-mediated suppression of ZO-3 resulted in increased cyclin D1 proteolysis and G(0)/G(1) cell-cycle retention. This study highlights an important new role for ZO family TJ proteins in regulating epithelial cell proliferation through stabilization of cyclin D1 during mitosis.
Sigma-Aldrich
Monoclonal Anti-Cyclin D1 antibody produced in mouse, clone DCS-6, ascites fluid