MilliporeSigma
  • Background-free Detection of Mouse Antibodies on Mouse Tissue by Anti-isotype Secondary Antibodies.
Background-free Detection of Mouse Antibodies on Mouse Tissue by Anti-isotype Secondary Antibodies.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2021-07-21)
Francesco Mascadri, Roberta Ciccimarra, Maddalena M Bolognesi, Fabio Stellari, Francesca Ravanetti, Giorgio Cattoretti
ABSTRACT
Immunodetection on mouse routinely processed tissue via antibodies raised in mice faces cross-reactivity of the secondary anti-mouse reagents with endogenous immunoglobulins, which permeate the tissue. Various solutions to this problem have been devised and include endogenous Ig block with anti-mouse Fab fragments or directly conjugated primary antibodies. Mouse isotype-specific antibodies, differently from reagents directed against both heavy and light chains, fail to detect endogenous Ig after fixation and embedding, while providing a clean and specific detection system for mouse antibodies on mouse routinely processed tissue.
Sigma-Aldrich
Anti-Mouse IgG2a (γ2a), CF488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution