MilliporeSigma
  • Killer cell lectin-like receptor G1 deficiency significantly enhances survival after Mycobacterium tuberculosis infection.
Killer cell lectin-like receptor G1 deficiency significantly enhances survival after Mycobacterium tuberculosis infection.
Infection and immunity (2013-01-24)
Joshua C Cyktor, Bridget Carruthers, Paul Stromberg, Emilio Flaño, Hanspeter Pircher, Joanne Turner
ABSTRACT
The expression of T cell differentiation markers is known to increase during Mycobacterium tuberculosis infection, and yet the biological role of such markers remains unclear. We examined the requirement of the T cell differentiation marker killer cell lectin-like receptor G1 (KLRG1) during M. tuberculosis infection using mice deficient in KLRG1. KLRG1(-/-) mice had a significant survival extension after M. tuberculosis infection compared to wild-type controls, and maintained a significantly lower level of pulmonary M. tuberculosis throughout chronic infection. Improved control of M. tuberculosis infection was associated with an increased number of activated pulmonary CD4(+) T cells capable of secreting gamma interferon (IFN-γ). Our report is the first to show an in vivo impact of KLRG1 on disease control.