MilliporeSigma
  • Nonvolatile bipolar resistive memory switching in single crystalline NiO heterostructured nanowires.
Nonvolatile bipolar resistive memory switching in single crystalline NiO heterostructured nanowires.
Journal of the American Chemical Society (2009-02-21)
Keisuke Oka, Takeshi Yanagida, Kazuki Nagashima, Hidekazu Tanaka, Tomoji Kawai
ABSTRACT
We have demonstrated the nonvolatile bipolar resistive memory switching in single crystalline NiO heterostructured nanowires for the first time. The self-assembled NiO nanowires are expected to open up opportunities to explore not only the detailed nanoscale mechanisms in NiO resistive memory switching but also next-generation nanoscale nonvolatile memory devices with the potential for high-density device integration and improved memory characteristics.
Sigma-Aldrich
Nickel(II) oxide, nanowires, diam. × L ~20 nm × 10 μm