MilliporeSigma
  • Assessment of pesticide residues in strawberries grown under various treatment regimes.
Assessment of pesticide residues in strawberries grown under various treatment regimes.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2013-12-07)
Jana Kovacova, Vojtech Hrbek, Jana Kloutvorova, Vladimir Kocourek, Lucie Drabova, Jana Hajslova
ABSTRACT
The dynamics of pesticide residues in strawberries that involved quantification of pesticide residues in ripe fruits after model treatment was evaluated in repeated field trials conducted over 3 years. Sixteen commercial pesticide formulations in various combinations were employed in applications from 7 to 44 days before harvest. Altogether 21 active ingredients and some of their metabolites were determined in treated strawberries using LC-MS and GC-MS methods. Except for propargite, the concentrations of all active ingredients declined below the respective MRLs (Regulation (EC) No. 396/2005); nevertheless, most of the tested fungicides often persisted above the 0.01 mg kg⁻¹ limit required by baby food producers to avoid the risk of exceeding the 'baby food limit' established in Commission Directive 2006/141/EC. On the other hand, residues of the majority of tested insecticides, namely spinosad, pymetrozine, deltamethrin, lambda-cyhalothrin and azadirachtin, declined below this limit.
Product Description
Supelco
Pymetrozin Metabolite CGA313124, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland