MilliporeSigma
  • Stabilization of membrane-associated alpha-L-fucosidase by the human sperm equatorial segment.
Stabilization of membrane-associated alpha-L-fucosidase by the human sperm equatorial segment.
International journal of andrology (2008-06-05)
Jennifer J Venditti, Barry S Bean
ABSTRACT
Previous reports from this laboratory documented the existence of two novel isoforms of alpha-L-fucosidase in human semen and showed that membrane-associated alpha-L-fucosidase is cryptically held within the acrosomal compartment and enriched within the sperm equatorial segment. The occurrence of these novel isoforms is provocative. Sperm proteins potentially involved in sperm-egg interactions must maintain their functional integrity as they travel through the female reproductive tract. The goal of this project was to investigate the stability of membrane-associated alpha-L-fucosidase in human sperm. Whole seminal plasma and Percoll purified sperm cell populations were incubated for 72 h at 37 degrees C, with 5% CO(2) or ambient air. At various times during prolonged incubation, sperm cells were permeabilized with 0.1% Triton X-100 and enzyme assays using the fluorogenic substrate 4-MU-fuc were performed to evaluate the stability of both the seminal plasma and membrane-associated alpha-L-fucosidase. Here, we report seminal plasma alpha-L-fucosidase activity rapidly decreased within 24 h. Conversely, alpha-L-fucosidase activity from Percoll purified sperm cell populations persisted up to 72 h. Data from these experiments support the notions that (i) membrane-associated alpha-L-fucosidase is stable for extended periods of time, consistent with a possible role in sperm-egg interaction and (ii) membrane domains and compartmentalization within the human sperm are key to preserving protein integrity.
Sigma-Aldrich
4-Methylumbelliferyl α-L-fucopyranoside, ≥97% (TLC)