MilliporeSigma
  • Increased systemic exposure of fimasartan, an angiotensin II receptor antagonist, by ketoconazole and rifampicin.
Increased systemic exposure of fimasartan, an angiotensin II receptor antagonist, by ketoconazole and rifampicin.
Journal of clinical pharmacology (2013-02-13)
Jung Won Kim, SoJeong Yi, Tae-Eun Kim, Kyoung Soo Lim, Seo Hyun Yoon, Joo-Youn Cho, Min Goo Lee, Im-Sook Song, Sang-Goo Shin, In-Jin Jang, Kyung-Sang Yu
ABSTRACT
The authors studied the effects of ketoconazole and rifampicin on the pharmacokinetics of a single dose of fimasartan (BR-A-657), a newly developed angiotensin II receptor antagonist for the treatment of hypertension, in 22 healthy participants. Ketoconazole increased the maximumplasma concentration (Cmax) and area under the plasma concentration vs time curve to infinity (AUC∞ of fimasartan by 2.47-fold (90% confidence interval [CI], 1.61-3.79) and 2.03-fold (1.56-2.64), respectively. Concomitant administration of rifampicin increased the C(max) and AUC∞ of fimasartan by 10.33-fold (90% CI, 6.74-15.81) and 4.60-fold (3.54-5.97). In vitro studies indicated that ketoconazole inhibited the uptake of fimasartan into cells expressing OATP1B1 with a K(i) of 107.7 µM, and rifampicin inhibited OAT1- and OATP1B1-mediated fimasartan transport with a K(i) of 212 µM and 12.2 µM, respectively. The systemic exposure of fimasartan was significantly increased by coadministration of ketoconazole or rifampicin in healthy volunteers. This is consistent with the in vitro results, in which fimasartan is a substrate of CYP3A and OATP1B1.
Product Description
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
Fimasartan, ≥98% (HPLC)