MilliporeSigma
  • Cardioprotective effect of fimasartan, a new angiotensin receptor blocker, in a porcine model of acute myocardial infarction.
Cardioprotective effect of fimasartan, a new angiotensin receptor blocker, in a porcine model of acute myocardial infarction.
Journal of Korean medical science (2015-01-02)
Doo Sun Sim, Myung Ho Jeong, Ho Chun Song, Jahae Kim, Ari Chong, Hee Seung Bom, In Seok Jeong, Sang Gi Oh, Jong Min Kim, Dae Sung Park, Jung Ha Kim, Kyung Seob Lim, Min Suk Kim, Shi Hyun Ryu, Hyun Kuk Kim, Sung Soo Kim, Su Young Jang, Jae Yeong Cho, Hae Chang Jeong, Ki Hong Lee, Keun Ho Park, Nam Sik Yoon, Hyun Ju Yoon, Kye Hun Kim, Young Joon Hong, Hyung Wook Park, Ju Han Kim, Youngkeun Ahn, Jeong Gwan Cho, Jong Chun Park, Jung Chaee Kang
ABSTRACT
Cardioprotective effect of fimasartan, a new angiotensin receptor blocker (ARB), was evaluated in a porcine model of acute myocardial infarction (MI). Fifty swine were randomized to group 1 (sham, n=10), group 2 (no angiotensin-converting enzyme inhibitor [ACEI] or ARB, n=10), group 3 (perindopril 2 mg daily, n=10), group 4 (valsartan 40 mg daily, n=10), or group 5 (fimasartan 30 mg daily, n=10). Acute MI was induced by occlusion of the left anterior descending artery for 50 min. Echocardiography, single photon emission computed tomography (SPECT), and F-18 fluorodeoxyglucose cardiac positron emission tomography (PET) were performed at baseline, 1 week, and 4 weeks. Iodine-123 meta-iodobenzylguanidine (MIBG) scan was done at 6 weeks for visualization of cardiac sympathetic activity. Left ventricular function and volumes at 4 weeks were similar between the 5 groups. No difference was observed in groups 2 to 5 in SPECT perfusion defect, matched and mismatched segments between SPECT and PET at 1 week and 4 weeks. MIBG scan showed similar uptake between the 5 groups. Pathologic analysis showed similar infarct size in groups 2 to 5. Infarct size reduction was not observed with use of fimasartan as well as other ACEI and ARB in a porcine model of acute MI.
Sigma-Aldrich
Fimasartan, ≥98% (HPLC)