MilliporeSigma
Müllerian remnants in complete androgen insensitivity syndrome.
International journal of fertility (1992-09-01)
D S Heller, A Ranzini, W Futterweit, P Dottino, L Deligdisch
ABSTRACT
Patients with complete androgen insensitivity syndrome rarely have Müllerian remnants. A patient with this syndrome found to have a microscopic fallopian tube at the time of gonadectomy is described and possible etiologies for the finding are discussed.
Sigma-Aldrich
PF74, ≥98% (HPLC)