MilliporeSigma
  • Efficacy and safety of telaprevir with natural human interferon-β and ribavirin in Japanese chronic hepatitis C patients with depression.
Efficacy and safety of telaprevir with natural human interferon-β and ribavirin in Japanese chronic hepatitis C patients with depression.
Hepatology research : the official journal of the Japan Society of Hepatology (2017-05-13)
Hiromitsu Kumada, Satoshi Mochida, Makoto Nakamuta, Fumitaka Suzuki, Takashi Yagi, Ryuji Takasaki, Masao Okai, Naohiro Kamiya, Yasushi Okada, Saya Hirota, Madori Orihashi, Miyoko Ochi, Kazuaki Chayama
ABSTRACT
To assess the efficacy and safety of telaprevir (TVR) when used in combination with natural human interferon-β (IFN-β) and ribavirin (RBV) for genotype 1 patients with depression compared to IFN-β/RBV therapy in Japan. We also examined the efficacy of the TVR/IFN-β/RBV therapy in treatment failure genotype 2 patients with depression. For the genotype 1 patients, 30 patients received TVR (750 mg every 8 h) for 12 weeks combined with IFN-β and RBV for 24 weeks (Group A), and 30 received IFN-β and RBV for 48 weeks (Group B). For the genotype 2 patients, 14 patients were dosed only with the TVR-based regimen. The sustained virologic response (SVR) rates for Group A and Group B were 63.3% and 20.0%, respectively (P = 0.001, likelihood ratio test). The SVR rate for genotype 2 patients previously treated with pegylated IFN and/or RBV was 71.4%. No patient dropped out due to exacerbation of depression. The trend of platelet counts after the drugs were given was similar in the TVR/IFN-β/RBV therapy group and the IFN-β/RBV therapy group. Common resistance-associated variants of TVR were identified in 4 of the 13 patients who did not achieve SVR. This study showed that an addition of TVR to IFN-β/RBV therapy raised SVR in previously treated and untreated genotype 1 patients and previously treated genotype 2 patients with chronic hepatitis C and depression.
Sigma-Aldrich
Telaprevir, ≥90% (HPLC)