MilliporeSigma
  • The Small Molecule BC-2059 Inhibits Wingless/Integrated (Wnt)-Dependent Gene Transcription in Cancer through Disruption of the Transducin β-Like 1-β-Catenin Protein Complex.
The Small Molecule BC-2059 Inhibits Wingless/Integrated (Wnt)-Dependent Gene Transcription in Cancer through Disruption of the Transducin β-Like 1-β-Catenin Protein Complex.
The Journal of pharmacology and experimental therapeutics (2021-05-20)
Raffaella Soldi, Tithi Ghosh Halder, Samuel Sampson, Hariprasad Vankayalapati, Alexis Weston, Trason Thode, Kapil N Bhalla, Serina Ng, Ryan Rodriguez Del Villar, Kevin Drenner, Mohan R Kaadige, Stephen K Horrigan, Surinder K Batra, Ravi Salgia, Sunil Sharma
ABSTRACT
The central role of β-catenin in the Wnt pathway makes it an attractive therapeutic target for cancers driven by aberrant Wnt signaling. We recently developed a small-molecule inhibitor, BC-2059, that promotes apoptosis by disrupting the β-catenin/transducin β-like 1 (TBL1) complex through an unknown mechanism of action. In this study, we show that BC-2059 directly interacts with high affinity for TBL1 when in complex with β-catenin. We identified two amino acids in a hydrophobic pocket of TBL1 that are required for binding with β-catenin, and computational modeling predicted that BC-2059 interacts at the same hydrophobic pocket. Although this pocket in TBL1 is involved in binding with NCoR/SMRT complex members G Protein Pathway Suppressor 2 (GSP2) and SMRT and p65 NFκB subunit, BC-2059 failed to disrupt the interaction of TBL1 with either NCoR/SMRT or NFκB. Together, our results show that BC-2059 selectively targets TBL1/β-catenin protein complex, suggesting BC-2059 as a therapeutic for tumors with deregulated Wnt signaling pathway. SIGNIFICANCE STATEMENT: This study reports the mechanism of action of a novel Wnt pathway inhibitor, characterizing the selective disruption of the transducin β-like 1/β-catenin protein complex. As Wnt signaling is dysregulated across cancer types, this study suggests BC-2059 has the potential to benefit patients with tumors reliant on this pathway.
Sigma-Aldrich
BC-2059, ≥98% (HPLC)