MilliporeSigma
  • Analysis of pesticide residues in commercially available chenpi using a modified QuEChERS method and GC-MS/MS determination.
Analysis of pesticide residues in commercially available chenpi using a modified QuEChERS method and GC-MS/MS determination.
Journal of pharmaceutical analysis (2020-03-04)
Shuang Li, Peipei Yu, Ceng Zhou, Ling Tong, Dongxiang Li, Zhiguo Yu, Yunli Zhao
ABSTRACT
To ensure the safety of the commercially available chenpi, a convenient and fast analytical method was developed for the determination of 133 pesticide residues in chenpi using gas chromatography-tandem mass spectrometry (GC-MS/MS). In this study, different extraction solvents, redissolution solvents and adsorbents were tested according to the recovery and purification effect to obtain a modified QuEChERS method. The samples were extracted with acetonitrile. During the clean-up step, octadecyl-modified silica (C18) and graphitized carbon black (GCB) were selected, and aminopropyl (NH2) was used instead of primary secondary amine (PSA) because of its weaker ion exchange capacity which had little effect on the recovery of ditalimfos. Samples were quantified by matrix-matched calibration with internal standards. All pesticides showed good linearity in the respective range, both with values of r 2 > 0.99. The average recoveries of the pesticides spiked samples ranged from 70.0% to 112.2% with the RSDs of 0.2%-14.4%. The modified QuEChERS method was validated and applied to twenty real samples. Five pesticides were found in eight batches, but no pesticide exceeded the maximum residue limits (MRL, MRL reference to European commission).
Product Description
Supelco
Prothiofos, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland